accessibility menu, dialog, popup

UserName

CAS RN 1192-28-5

IUPAC Name: N-cyclopentylidenehydroxylamine
Synonyms: cyclopentanone oxime
Molecular Weight (g/mol): 99.13
Molecular Formula: C5H9NO
InChi Key: YGNXYFLJZILPEK-UHFFFAOYSA-N
SMILES: ON=C1CCCC1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)