accessibility menu, dialog, popup

UserName

CAS RN 119192-09-5

NNNNOO

CAS RN 119192-09-5

IUPAC Name: 1-[(4-nitrophenyl)methyl]-1H-1,2,4-triazole
Synonyms: 1-[(4-nitrophenyl)methyl]-1,2,4-triazole
Molecular Weight (g/mol): 204.19
Molecular Formula: C9H8N4O2
InChi Key: NVRYCUYVBBCXHT-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(CN2C=NC=N2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results