accessibility menu, dialog, popup

UserName

CAS RN 119110-38-2

IUPAC Name: N-heptyl-N'-(8-{2-[heptyl(methyl)carbamoyl]acetamido}octyl)-N-methylpropanediamide
Synonyms: N-heptyl-N'-(8-{2-[heptyl(methyl)carbamoyl]acetamido}octyl)-N-methylpropanediamide
Molecular Weight (g/mol): 538.82
Molecular Formula: C30H58N4O4
InChi Key: MXMJPDBVBJZBCA-UHFFFAOYSA-N
SMILES: CCCCCCCN(C)C(=O)CC(=O)NCCCCCCCCNC(=O)CC(=O)N(C)CCCCCCC
Grade 
  • (1)
Product Line 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results