accessibility menu, dialog, popup

UserName

Check Out the New Spotlight Offers and Get Up to 50% Off

Save Today

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 1190-39-2

H3COOOOCH3

CAS RN 1190-39-2

IUPAC Name: 1,3-dibutyl propanedioate
Synonyms: propanedioic acid, dibutyl ester
Molecular Weight (g/mol): 216.28
Molecular Formula: C11H20O4
InChi Key: NFKGQHYUYGYHIS-UHFFFAOYSA-N
SMILES: CCCCOC(=O)CC(=O)OCCCC
Boiling Point 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.