accessibility menu, dialog, popup

UserName

H2NNHNOONOO

CAS RN 119-26-6

IUPAC Name: (2,4-dinitrophenyl)hydrazine
Synonyms: 2,4-dinitrophenylhydrazine
Molecular Weight (g/mol): 198.14
Molecular Formula: C6H6N4O4
InChi Key: HORQAOAYAYGIBM-UHFFFAOYSA-N
SMILES: NNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Physical Form 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (4)
Percent Purity 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)