accessibility menu, dialog, popup

UserName

H3COONOHO

CAS RN 118133-15-6

IUPAC Name: 1-(ethoxycarbonyl)piperidine-4-carboxylic acid
Synonyms: 1-(ethoxycarbonyl)piperidine-4-carboxylic acid
Molecular Weight (g/mol): 201.22
Molecular Formula: C9H15NO4
InChi Key: BVEBMPFJSMEGJX-UHFFFAOYSA-N
SMILES: CCOC(=O)N1CCC(CC1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results