CAS RN 117552-78-0
IUPAC Name:
calcium bis(2,5-dihydroxybenzene-1-sulfonate) hydrate
Synonyms:
calcium bis(2,5-dihydroxybenzenesulfonate) hydrate
Molecular Weight (g/mol):
436.42
Molecular Formula:
C12H12CaO11S2
InChi Key:
JMTQBGBMQZPWCS-UHFFFAOYSA-L
SMILES:
O.[Ca++].OC1=CC(=C(O)C=C1)S([O-])(=O)=O.OC1=CC(=C(O)C=C1)S([O-])(=O)=O
1
–
1
of
1
results
1
Calcium Dobesilate Hydrate 98.0+%, TCI America™
CAS: 117552-78-0 Molecular Formula: C12H12CaO11S2 Molecular Weight (g/mol): 436.415 MDL Number: MFCD00867249 InChI Key: JMTQBGBMQZPWCS-UHFFFAOYSA-L Synonym: Calcium 2,5-Dihydroxybenzenesulfonate, 2,5-Dihydroxybenzenesulfonic Acid Calcium Salt PubChem CID: 45358052 IUPAC Name: calcium;2,5-dihydroxybenzenesulfonate;hydrate SMILES: C1=CC(=C(C=C1O)S(=O)(=O)[O-])O.C1=CC(=C(C=C1O)S(=O)(=O)[O-])O.O.[Ca+2]