accessibility menu, dialog, popup

UserName

CAS RN 116855-03-9

H3CNNNBr

CAS RN 116855-03-9

IUPAC Name: 3-bromo-1-methyl-1H-pyrazolo[3,4-b]pyridine
Synonyms: 3-bromo-1-methylpyrazolo[3,4-b]pyridine
Molecular Weight (g/mol): 212.05
Molecular Formula: C7H6BrN3
InChi Key: BVDWIHGOGVWCCJ-UHFFFAOYSA-N
SMILES: CN1N=C(Br)C2=CC=CN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)