CAS RN 116153-81-2
IUPAC Name:
5-(furan-2-yl)-1H-pyrazole-3-carboxylic acid
Synonyms:
5-(furan-2-yl)-1H-pyrazole-3-carboxylic acid
Molecular Weight (g/mol):
178.15
Molecular Formula:
C8H6N2O3
InChi Key:
GKPSFQIKCROJOB-UHFFFAOYSA-N
SMILES:
OC(=O)C1=NNC(=C1)C1=CC=CO1
1
–
1
of
1
results
1
5-(2-Furyl)-1H-pyrazole-3-carboxylic acid, 97%, Thermo Scientific™
CAS: 116153-81-2 Molecular Formula: C8H6N2O3 Molecular Weight (g/mol): 178.147 MDL Number: MFCD05170057 InChI Key: GKPSFQIKCROJOB-UHFFFAOYSA-N Synonym: 5-2-furyl-1h-pyrazole-3-carboxylic acid,5-furan-2-yl-1h-pyrazole-3-carboxylic acid,5-furan-2-yl-2h-pyrazole-3-carboxylic acid,3-furan-2-yl-1h-pyrazole-5-carboxylic acid,3-2-furyl-1h-pyrazole-5-carboxylic acid,1h-pyrazole-3-carboxylicacid, 5-2-furanyl,5-2-furyl pyrazole-3-carboxylic acid,acmc-20f1sn,3-2-furyl pyrazole-5-carboxylic acid PubChem CID: 654018 IUPAC Name: 5-(furan-2-yl)-1H-pyrazole-3-carboxylic acid SMILES: C1=COC(=C1)C2=CC(=NN2)C(=O)O