CAS RN 1160102-28-2
IUPAC Name:
6-bromo-7-fluoro-3,4-dihydro-2H-1,4-benzoxazine
Synonyms:
6-bromo-7-fluoro-3,4-dihydro-2H-1,4-benzoxazine
Molecular Weight (g/mol):
232.05
Molecular Formula:
C8H7BrFNO
InChi Key:
CTMRDIDZVAZIRX-UHFFFAOYSA-N
SMILES:
FC1=C(Br)C=C2NCCOC2=C1
1
–
1
of
1
results
1
Promotions Available
6-Bromo-7-fluoro-3,4-dihydro-2H-1,4-benzoxazine, 96%, Thermo Scientific Chemicals
6-Bromo-7-fluoro-3,4-dihydro-2H-1,4-benzoxazine, 96%, Thermo Scientific Chemicals
CAS: 1160102-28-2 Molecular Formula: C8H7BrFNO Molecular Weight (g/mol): 232.05 MDL Number: MFCD20441785 InChI Key: CTMRDIDZVAZIRX-UHFFFAOYSA-N Synonym: 6-bromo-7-fluoro-3,4-dihydro-2h-benzo b 1,4 oxazine,acmc-2099rp PubChem CID: 56776541 IUPAC Name: 6-bromo-7-fluoro-3,4-dihydro-2H-1,4-benzoxazine SMILES: FC1=C(Br)C=C2NCCOC2=C1