accessibility menu, dialog, popup

UserName

CH3HNOONH3CCH3SCH3(E)

CAS RN 116-06-3

IUPAC Name: (E)-[2-methyl-2-(methylsulfanyl)propylidene]amino N-methylcarbamate
Synonyms: temik rcra waste number P070 temik G10 temik G aldicarb (substance) temik 10 G union carbide UC-21149 sulfone aldoxycarb union carbide 21149 ccris 17
Molecular Weight (g/mol): 190.26
Molecular Formula: C7H14N2O2S
InChi Key: QGLZXHRNAYXIBU-WEVVVXLNSA-N
SMILES: CNC(=O)O\N=C\C(C)(C)SC
Grade 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)