CAS RN 115532-50-8
IUPAC Name:
6-(dimethylamino)-9-[2-(methoxycarbonyl)phenyl]-N,N-dimethyl-3H-xanthen-3-iminium perchlorate
Synonyms:
6-(dimethylamino)-9-[2-(methoxycarbonyl)phenyl]-N,N-dimethylxanthen-3-iminium perchlorate
Molecular Weight (g/mol):
500.93
Molecular Formula:
C25H25ClN2O7
InChi Key:
PFYWPQMAWCYNGW-UHFFFAOYSA-M
SMILES:
[O-][Cl](=O)(=O)=O.COC(=O)C1=CC=CC=C1C1=C2C=CC(C=C2OC2=CC(=CC=C12)N(C)C)=[N+](C)C
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Tetramethylrhodamine methyl ester perchlorate
Thermo Scientific Chemicals Tetramethylrhodamine methyl ester perchlorate
CAS: 115532-50-8 Molecular Formula: C25H25ClN2O7 Molecular Weight (g/mol): 500.93 InChI Key: PFYWPQMAWCYNGW-UHFFFAOYSA-M IUPAC Name: 6-(dimethylamino)-9-[2-(methoxycarbonyl)phenyl]-N,N-dimethyl-3H-xanthen-3-iminium perchlorate SMILES: [O-][Cl](=O)(=O)=O.COC(=O)C1=CC=CC=C1C1=C2C=CC(C=C2OC2=CC(=CC=C12)N(C)C)=[N+](C)C