missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-2-ethyl-1,3-propanediol, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| CAS | 115-70-8 |
| CAS Min % | 96.0 |
| CAS Max % | 100.0 |
| Density | 1.0990g/mL |
| Boiling Point | 152°C to 153°C (10.0mmHg) |
| Flash Point | >110°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97% |
| Packaging | Glass bottle |
| Molecular Formula | C5H13NO2 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes serious eye damage. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doc |
| EINECSNumber | 204-101-2 |
| TSCA | TSCA |
RUO – Research Use Only