accessibility menu, dialog, popup

UserName

CAS RN 114622-81-0

IUPAC Name: (2S)-5-{[amino(iminiumyl)methyl]amino}-2-{[(tert-butoxy)carbonyl]amino}pentanoate
Synonyms: (2S)-5-{[amino(iminio)methyl]amino}-2-[(tert-butoxycarbonyl)amino]pentanoate
Molecular Weight (g/mol): 274.32
Molecular Formula: C11H22N4O4
InChi Key: HSQIYOPBCOPMSS-ZETCQYMHSA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CCCNC(N)=[NH2+])C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results