accessibility menu, dialog, popup

UserName

CAS RN 1145-73-9

H3CNCH3(E)

CAS RN 1145-73-9

IUPAC Name: N,N-dimethyl-4-[(1E)-2-phenylethenyl]aniline
Synonyms: 4-dimethylaminostilbene
Molecular Weight (g/mol): 223.32
Molecular Formula: C16H17N
InChi Key: XGHHHPDRXLIMPM-CMDGGOBGSA-N
SMILES: CN(C)C1=CC=C(\C=C\C2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results