CAS RN 1144573-61-4
IUPAC Name:
potassium trifluoro(6-methoxypyridin-3-yl)boranuide
Synonyms:
potassium trifluoro(6-methoxypyridin-3-yl)boranuide
Molecular Weight (g/mol):
215.02
Molecular Formula:
C6H6BF3KNO
InChi Key:
ASWXYOAUUZSFCK-UHFFFAOYSA-N
SMILES:
[K+].COC1=NC=C(C=C1)[B-](F)(F)F
1
–
1
of
1
results
1
Promotions Available
Potassium 2-methoxypyridine-5-trifluoroborate, 97%, Thermo Scientific™
Potassium 2-methoxypyridine-5-trifluoroborate, 97%, Thermo Scientific™
CAS: 1144573-61-4 Molecular Formula: C6H6BF3KNO Molecular Weight (g/mol): 215.024 MDL Number: MFCD09993111 InChI Key: ASWXYOAUUZSFCK-UHFFFAOYSA-N Synonym: potassium 2-methoxypyridine-5-trifluoroborate,potassium trifluoro 6-methoxypyridin-3-yl borate,amtb168,potassium 6-methoxy-3-pyridyltrifluoroborate,potassium trifluoro 6-methoxypyridin-3-yl boranuide PubChem CID: 72698470 IUPAC Name: potassium;trifluoro-(6-methoxypyridin-3-yl)boranuide SMILES: [B-](C1=CN=C(C=C1)OC)(F)(F)F.[K+]