accessibility menu, dialog, popup

UserName

OHONHONN

CAS RN 114457-94-2

IUPAC Name: 3-phenyl-2-[(pyrazin-2-yl)formamido]propanoic acid
Synonyms: 3-phenyl-2-(pyrazin-2-ylformamido)propanoic acid
Molecular Weight (g/mol): 271.28
Molecular Formula: C14H13N3O3
InChi Key: DWYZPDHMMZGQAP-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=CC=C1)NC(=O)C1=NC=CN=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results