CAS RN 1127-08-8
IUPAC Name:
cyclohexane-1,1-dicarboxylic acid
Synonyms:
cyclohexane-1,1-dicarboxylic acid
Molecular Weight (g/mol):
172.18
Molecular Formula:
C8H12O4
InChi Key:
QYQADNCHXSEGJT-UHFFFAOYSA-N
SMILES:
OC(=O)C1(CCCCC1)C(O)=O
1
–
1
of
1
results
1
Promotions Available
1,1-Cyclohexanedicarboxylic acid, 95%
1,1-Cyclohexanedicarboxylic acid, 95%
CAS: 1127-08-8 Molecular Formula: C8H12O4 Molecular Weight (g/mol): 172.18 MDL Number: MFCD00094668 InChI Key: QYQADNCHXSEGJT-UHFFFAOYSA-N Synonym: cyclohexanedicarboxylic acid,1,1-cyclohexanedicarboxylic acid,acmc-20ald2,cyclohexane dicarboxylic acid,1,1-cyclohexanedicarboxylicacid,cyclohexane-1,1-dicarboxylicacid,cyclohexane-1,1-dicarboxylic acid PubChem CID: 136906 IUPAC Name: cyclohexane-1,1-dicarboxylic acid SMILES: C1CCC(CC1)(C(=O)O)C(=O)O