accessibility menu, dialog, popup

UserName

CH3SNNOHO

CAS RN 1126-44-9

IUPAC Name: 2-(methylsulfanyl)pyrimidine-4-carboxylic acid
Synonyms: 2-(methylsulfanyl)pyrimidine-4-carboxylic acid
Molecular Weight (g/mol): 170.19
Molecular Formula: C6H6N2O2S
InChi Key: IAGNLKODEFUQDV-UHFFFAOYSA-N
SMILES: CSC1=NC=CC(=N1)C(O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results