accessibility menu, dialog, popup

UserName

H3CNCH3Cl

CAS RN 112400-86-9

IUPAC Name: 1-butyl-4-methylpyridin-1-ium chloride
Synonyms: 1-butyl-4-methylpyridin-1-ium chloride
Molecular Weight (g/mol): 185.7
Molecular Formula: C10H16ClN
InChi Key: YTSDTJNDMGOTFN-UHFFFAOYSA-M
SMILES: [Cl-].CCCC[N+]1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results