CAS RN 1123-40-6
IUPAC Name:
4,4-dimethylpiperidine-2,6-dione
Synonyms:
glutarimide, 3,3-dimethyl-
Molecular Weight (g/mol):
141.17
Molecular Formula:
C7H11NO2
InChi Key:
YUJCWMGBRDBPDL-UHFFFAOYSA-N
SMILES:
CC1(C)CC(=O)NC(=O)C1
1
–
1
of
1
results
1
3,3-Dimethylglutarimide 95.0+%, TCI America™
CAS: 1123-40-6 Molecular Formula: C7H11NO2 Molecular Weight (g/mol): 141.17 MDL Number: MFCD00006671 InChI Key: YUJCWMGBRDBPDL-UHFFFAOYSA-N Synonym: 3,3-dimethylglutarimide,2,6-piperidinedione, 4,4-dimethyl,glutarimide, 3,3-dimethyl,4,4-dimethyl-2,6-piperidinedione,beta,beta-dimethylglutarimide,3,3-dimethyl-glutarimide,.beta.,.beta.-dimethylglutarimide,4,6-piperidinedione,2, 4,4-dimethyl,glutarimide,3-dimethyl PubChem CID: 14292 IUPAC Name: 4,4-dimethylpiperidine-2,6-dione SMILES: CC1(CC(=O)NC(=O)C1)C