accessibility menu, dialog, popup

UserName

BrHOHOBr(?)

CAS RN 111795-43-8

IUPAC Name: 3,3'-dibromo-[1,1'-binaphthalene]-2,2'-diol
Synonyms: 3,3'-dibromo-[1,1'-binaphthalene]-2,2'-diol
Molecular Weight (g/mol): 444.12
Molecular Formula: C20H12Br2O2
InChi Key: BRTBEAXHUYEXSY-UHFFFAOYSA-N
SMILES: OC1=C(C2=CC=CC=C2C=C1Br)C1=C2C=CC=CC2=CC(Br)=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results