accessibility menu, dialog, popup

UserName

H3CNH2HCl

CAS RN 111180-78-0

IUPAC Name: 2-methylnaphthalen-1-amine hydrochloride
Synonyms: 1-naphthylamine, 2-methyl- hydrochloride
Molecular Weight (g/mol): 193.67
Molecular Formula: C11H12ClN
InChi Key: PEBKGVSIRFSIGW-UHFFFAOYSA-N
SMILES: Cl.CC1=CC=C2C=CC=CC2=C1N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results