accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNHNCH3CH3

CAS RN 1111637-92-3

IUPAC Name: N,3-dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Synonyms: N,3-dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Molecular Weight (g/mol): 248.13
Molecular Formula: C13H21BN2O2
InChi Key: VZNAKHVRKWXDMU-UHFFFAOYSA-N
SMILES: CNC1=C(C)C=C(C=N1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results