accessibility menu, dialog, popup

UserName

FFOOOH

CAS RN 110931-77-6

IUPAC Name: 4-(2,4-difluorophenyl)-4-oxobutanoic acid
Synonyms: 4-(2,4-difluorophenyl)-4-oxobutanoic acid
Molecular Weight (g/mol): 214.17
Molecular Formula: C10H8F2O3
InChi Key: OKYUHFSCTFNDFB-UHFFFAOYSA-N
SMILES: OC(=O)CCC(=O)C1=C(F)C=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results