accessibility menu, dialog, popup

UserName

CAS RN 1100598-50-2

IUPAC Name: 5-bromo-2-[3-(chloromethyl)phenyl]pyrimidine
Synonyms: 5-bromo-2-[3-(chloromethyl)phenyl]pyrimidine
Molecular Weight (g/mol): 283.55
Molecular Formula: C11H8BrClN2
InChi Key: KHSZEUJOVUJWKR-UHFFFAOYSA-N
SMILES: ClCC1=CC=CC(=C1)C1=NC=C(Br)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results