accessibility menu, dialog, popup

UserName

H3CSOH

CAS RN 110-77-0

IUPAC Name: 2-(ethylsulfanyl)ethan-1-ol
Synonyms: 2-(ethylthio)ethanol
Molecular Weight (g/mol): 106.18
Molecular Formula: C4H10OS
InChi Key: LNRIEBFNWGMXKP-UHFFFAOYSA-N
SMILES: CCSCCO
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)