accessibility menu, dialog, popup

UserName

CAS RN 1093077-77-0

IUPAC Name: 1,2,4,5,6-pentakis[(11-{4-[2-(4-hexylphenyl)diazen-1-yl]phenoxy}undecanoyl)oxy]hexan-3-yl 11-{4-[2-(4-hexylphenyl)diazen-1-yl]phenoxy}undecanoate
Synonyms: 1,2,4,5,6-pentakis[(11-{4-[2-(4-hexylphenyl)diazen-1-yl]phenoxy}undecanoyl)oxy]hexan-3-yl 11-{4-[2-(4-hexylphenyl)diazen-1-yl]phenoxy}undecanoate
Molecular Weight (g/mol): 2874.08
Molecular Formula: C180H254N12O18
InChi Key: DMCHOYXACRXKRI-UHFFFAOYNA-N
SMILES: CCCCCCC1=CC=C(C=C1)N=NC1=CC=C(OCCCCCCCCCCC(=O)OCC(OC(=O)CCCCCCCCCCOC2=CC=C(C=C2)N=NC2=CC=C(CCCCCC)C=C2)C(OC(=O)CCCCCCCCCCOC2=CC=C(C=C2)N=NC2=CC=C(CCCCCC)C=C2)C(OC(=O)CCCCCCCCCCOC2=CC=C(C=C2)N=NC2=CC=C(CCCCCC)C=C2)C(COC(=O)CCCCCCCCCCOC2=CC=C(C=C2)N=NC2=CC=C(CCCCCC)C=C2)OC(=O)CCCCCCCCCCOC2=CC=C(C=C2)N=NC2=CC=C(CCCCCC)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results