accessibility menu, dialog, popup

UserName

CAS RN 108354-78-5

IUPAC Name: methyl 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Synonyms: methyl 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Molecular Weight (g/mol): 211.28
Molecular Formula: C10H13NO2S
InChi Key: DKYYKIHEIOOWRB-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)SC2=C1CCCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results