accessibility menu, dialog, popup

UserName

CAS RN 108320-87-2

IUPAC Name: disodium [5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl phosphonato]oxy}phosphonate
Synonyms: disodium [5-(2,4-dioxo-3H-pyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl [3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl phosphonato]oxyphosphonate
Molecular Weight (g/mol): 651.32
Molecular Formula: C17H25N3Na2O17P2
InChi Key: HXWKMJZFIJNGES-UHFFFAOYNA-L
SMILES: [Na+].[Na+].CC(=O)NC1C(O)C(O)C(CO)OC1OP([O-])(=O)OP([O-])(=O)OCC1OC(C(O)C1O)N1C=CC(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results