accessibility menu, dialog, popup

UserName

NOSOOFFF

CAS RN 107658-27-5

IUPAC Name: pyridin-3-yl trifluoromethanesulfonate
Synonyms: pyridin-3-yl trifluoromethanesulfonate
Molecular Weight (g/mol): 227.16
Molecular Formula: C6H4F3NO3S
InChi Key: ZRNDSLLIAOLRJS-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OC1=CN=CC=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results