CAS RN 107549-26-8
IUPAC Name:
bromo[(2,6-difluorophenyl)methyl]magnesium
Synonyms:
bromo[(2,6-difluorophenyl)methyl]magnesium
Molecular Weight (g/mol):
231.32
Molecular Formula:
C7H5BrF2Mg
InChi Key:
KEOOIBWMYNKEEF-UHFFFAOYSA-M
SMILES:
FC1=CC=CC(F)=C1C[Mg]Br
1
–
1
of
1
results
1
Promotions Available
2,6-Difluorobenzylmagnesium bromide, 0.25M in 2-MeTHF
2,6-Difluorobenzylmagnesium bromide, 0.25M in 2-MeTHF
CAS: 107549-26-8 Molecular Formula: C7H5BrF2Mg Molecular Weight (g/mol): 231.32 MDL Number: MFCD11113508 InChI Key: KEOOIBWMYNKEEF-UHFFFAOYSA-M Synonym: 2,6-difluorobenzylmagnesium bromide, 0.25m in 2-methf,2,6-difluorobenzylmagnesium bromide,grignard reagent PubChem CID: 73995681 IUPAC Name: bromo[(2,6-difluorophenyl)methyl]magnesium SMILES: FC1=CC=CC(F)=C1C[Mg]Br