CAS RN 1074-55-1
IUPAC Name:
1-methyl-4-propylbenzene
Synonyms:
1-methyl-4-propylbenzene
Molecular Weight (g/mol):
134.22
Molecular Formula:
C10H14
InChi Key:
JXFVMNFKABWTHD-UHFFFAOYSA-N
SMILES:
CCCC1=CC=C(C)C=C1
1
–
1
of
1
results
1
4-Propyltoluene 99.0+%, TCI America™
CAS: 1074-55-1 Molecular Formula: C10H14 Molecular Weight (g/mol): 134.222 MDL Number: MFCD00048698 InChI Key: JXFVMNFKABWTHD-UHFFFAOYSA-N Synonym: 1-Methyl-4-propylbenzene PubChem CID: 14095 IUPAC Name: 1-methyl-4-propylbenzene SMILES: CCCC1=CC=C(C=C1)C