accessibility menu, dialog, popup

UserName

H3COCH3O

CAS RN 1072-33-9

IUPAC Name: tridecyl acetate
Synonyms: tridecyl acetate
Molecular Weight (g/mol): 242.4
Molecular Formula: C15H30O2
InChi Key: ZDRNMODJXFOYMN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCOC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results