CAS RN 107-30-2
IUPAC Name:
chloro(methoxy)methane
Synonyms:
chloromethyl methyl ether
Molecular Weight (g/mol):
80.51
Molecular Formula:
C2H5ClO
InChi Key:
XJUZRXYOEPSWMB-UHFFFAOYSA-N
SMILES:
COCCl
1
–
1
of
1
results
1
Chloromethyl Methyl Ether Solution, 1000μg/mL in Methanol, Ultra Scientific
CAS: 107-30-2 Molecular Formula: C2H5ClO Molecular Weight (g/mol): 80.511 InChI Key: XJUZRXYOEPSWMB-UHFFFAOYSA-N PubChem CID: 7864 ChEBI: CHEBI:82271 IUPAC Name: chloro(methoxy)methane SMILES: COCCl