accessibility menu, dialog, popup

UserName

CAS RN 106867-97-4

IUPAC Name: 1-{[4-({[4,4'-bipyridin]-1-ium-1-yl}methyl)phenyl]methyl}-[4,4'-bipyridin]-1-ium dibromide
Synonyms: 1-[(4-{[4,4'-bipyridin]-1-ium-1-ylmethyl}phenyl)methyl]-[4,4'-bipyridin]-1-ium dibromide
Molecular Weight (g/mol): 576.34
Molecular Formula: C28H24Br2N4
InChi Key: QSNYIUIZTNLTGX-UHFFFAOYSA-L
SMILES: [Br-].[Br-].C(C1=CC=C(C[N+]2=CC=C(C=C2)C2=CC=NC=C2)C=C1)[N+]1=CC=C(C=C1)C1=CC=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results