accessibility menu, dialog, popup

UserName

CAS RN 106-90-1

H2COOO

CAS RN 106-90-1

IUPAC Name: (oxiran-2-yl)methyl prop-2-enoate
Synonyms: glycidyl acrylate
Molecular Weight (g/mol): 128.13
Molecular Formula: C6H8O3
InChi Key: RPQRDASANLAFCM-UHFFFAOYNA-N
SMILES: C=CC(=O)OCC1CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results