missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl Acrylate (stabilized with MEHQ) 95.0+%, TCI America™

Catalog No. G04975G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G04975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G04975G Supplier TCI America Supplier No. G04975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 106-90-1
Color Yellow
Boiling Point 65°C
Molecular Formula C6H8O3
Quantity 5 g
MDL Number MFCD00005138
UN Number 2929
Synonym Acrylic Acid Glycidyl Ester
InChI Key RPQRDASANLAFCM-UHFFFAOYSA-N
SMILES C=CC(=O)OCC1CO1
IUPAC Name oxiran-2-ylmethyl prop-2-enoate
Molecular Weight (g/mol) 128.127
PubChem CID 7836
Formula Weight 128.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Glycidyl Acrylate (stabilized with MEHQ)
Show More Show Less
EINECSNumber (2)-1000
RTECSNumber AS9275000
TSCA Yes
Recommended Storage Refrigerator