accessibility menu, dialog, popup

UserName

CAS RN 10574-69-3

OSSN

CAS RN 10574-69-3

IUPAC Name: 3-benzyl-2-sulfanylidene-1,3-thiazolidin-4-one
Synonyms: 3-benzyl-2-sulfanylidene-1,3-thiazolidin-4-one
Molecular Weight (g/mol): 223.31
Molecular Formula: C10H9NOS2
InChi Key: ZFHVUMCTGGAWBM-UHFFFAOYSA-N
SMILES: O=C1CSC(=S)N1CC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results