CAS RN 105316-98-1
IUPAC Name:
5-(2-bromoacetyl)-2,3-dihydro-1H-indol-2-one
Synonyms:
5-(2-bromoacetyl)-1,3-dihydroindol-2-one
Molecular Weight (g/mol):
254.08
Molecular Formula:
C10H8BrNO2
InChi Key:
WHLZVVMOQHTDAX-UHFFFAOYSA-N
SMILES:
BrCC(=O)C1=CC2=C(NC(=O)C2)C=C1
1
–
2
of
2
results
2
5-(Bromoacetyl)-2-oxoindoline, 97%, Thermo Scientific™
CAS: 105316-98-1 Molecular Formula: C10H8BrNO2 Molecular Weight (g/mol): 254.083 MDL Number: MFCD08690294 InChI Key: WHLZVVMOQHTDAX-UHFFFAOYSA-N Synonym: 5-bromoacetyl-2-oxoindoline,5-2-bromoacetyl indolin-2-one,5-bromoacetyl-1,3-dihydro-2h-indol-2-one,5-2-bromoacetyl-1,3-dihydroindol-2-one,2h-indol-2-one,5-2-bromoacetyl-1,3-dihydro,5-bromoacetyloxindole,acmc-20a59p,5-alpha-bromoacetyl oxindole,5-2-bromoacetyl-2-indolinone,2h-indol-2-one, 5-bromoacetyl-1,3-dihydro PubChem CID: 22099309 IUPAC Name: 5-(2-bromoacetyl)-1,3-dihydroindol-2-one SMILES: C1C2=C(C=CC(=C2)C(=O)CBr)NC1=O