accessibility menu, dialog, popup

UserName

CAS RN 10526-80-4

IUPAC Name: 2-(phosphonooxy)prop-2-enoic acid; cyclohexanamine
Synonyms: cyclohexylamine; phosphoenolpyruvic acid
Molecular Weight (g/mol): 267.22
Molecular Formula: C9H18NO6P
InChi Key: VHFCNZDHPABZJO-UHFFFAOYSA-N
SMILES: NC1CCCCC1.OC(=O)C(=C)OP(O)(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)