accessibility menu, dialog, popup

UserName

OO

CAS RN 10519-12-7

IUPAC Name: decahydronaphthalen-2-yl formate
Synonyms: decahydro-β-naphthyl formate
Molecular Weight (g/mol): 182.26
Molecular Formula: C11H18O2
InChi Key: NMFWOBQDDWFAAB-UHFFFAOYNA-N
SMILES: O=COC1CCC2CCCCC2C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results