accessibility menu, dialog, popup

UserName

H3COOOOCH3

CAS RN 104954-58-7

IUPAC Name: methyl 6-(acetyloxy)hexanoate
Synonyms: methyl 6-(acetyloxy)hexanoate
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: MFIRVMDIUPRJDB-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCOC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results