accessibility menu, dialog, popup

UserName

A(a)

CAS RN 104219-63-8

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: —
InChi Key: —
SMILES: —
Material 
  • (2)
Particle Size 
  • (2)
Quantity 
  • (1)
  • (1)
Technique 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)