accessibility menu, dialog, popup

UserName

FFFHNF

CAS RN 1041583-33-8

IUPAC Name: N-[(4-fluorophenyl)methyl]-2-(trifluoromethyl)aniline
Synonyms: N-[(4-fluorophenyl)methyl]-2-(trifluoromethyl)aniline
Molecular Weight (g/mol): 269.24
Molecular Formula: C14H11F4N
InChi Key: JCWUQELJSDRKEP-UHFFFAOYSA-N
SMILES: FC1=CC=C(CNC2=CC=CC=C2C(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results