CAS RN 104-90-5
IUPAC Name:
5-ethyl-2-methylpyridine
Synonyms:
5-ethyl-2-methylpyridine
Molecular Weight (g/mol):
121.18
Molecular Formula:
C8H11N
InChi Key:
NTSLROIKFLNUIJ-UHFFFAOYSA-N
SMILES:
CCC1=CC=C(C)N=C1
1
–
1
of
1
results
1
5-Ethyl-2-methylpyridine 95.0+%, TCI America™
CAS: 104-90-5 Molecular Formula: C8H11N Molecular Weight (g/mol): 121.18 MDL Number: MFCD00006344 InChI Key: NTSLROIKFLNUIJ-UHFFFAOYSA-N Synonym: aldehydine,5-ethyl-2-picoline,2-methyl-5-ethylpyridine,aldehydkollidin,aldehydecollidine,pyridine, 5-ethyl-2-methyl,2,5-aldehydine,2-picoline, 5-ethyl,3-ethyl-6-methylpyridine,6-methyl-3-ethylpyridine PubChem CID: 7728 IUPAC Name: 5-ethyl-2-methylpyridine SMILES: CCC1=CC=C(C)N=C1