accessibility menu, dialog, popup

UserName

CAS RN 1034-10-2

H2NOHOHOO

CAS RN 1034-10-2

IUPAC Name: 2-amino-3-[4-(4-hydroxyphenoxy)phenyl]propanoic acid
Synonyms: thyronine
Molecular Weight (g/mol): 273.29
Molecular Formula: C15H15NO4
InChi Key: KKCIOUWDFWQUBT-UHFFFAOYNA-N
SMILES: NC(CC1=CC=C(OC2=CC=C(O)C=C2)C=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results