accessibility menu, dialog, popup

UserName

CAS RN 10333-92-3

IUPAC Name: bis(2-amino-6,7-dihydro-3H-purin-6-one); sulfuric acid
Synonyms: bis(2-aminohypoxanthine); sulfuric acid
Molecular Weight (g/mol): 400.33
Molecular Formula: C10H12N10O6S
InChi Key: JXUOOUXZIHGPTG-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC1=NC(=O)C2=C(N1)N=CN2.NC1=NC(=O)C2=C(N1)N=CN2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)