accessibility menu, dialog, popup

UserName

CAS RN 10312-55-7

IUPAC Name: 2-aminobenzene-1,4-dicarboxylate
Synonyms: 2-aminobenzene-1,4-dicarboxylate
Molecular Weight (g/mol): 179.13
Molecular Formula: C8H5NO4
InChi Key: GPNNOCMCNFXRAO-UHFFFAOYSA-L
SMILES: NC1=CC(=CC=C1C([O-])=O)C([O-])=O
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)