accessibility menu, dialog, popup

UserName

CAS RN 10294-42-5

IUPAC Name: cerium(3+) tetrahydrate hydrogen sulfate sulfate
Synonyms: cerium(3+) tetrahydrate hydrogen sulfate sulfate
Molecular Weight (g/mol): 405.3
Molecular Formula: CeH9O12S2
InChi Key: ZBNJQJDKRZKDSV-UHFFFAOYSA-K
SMILES: O.O.O.O.[Ce+3].OS([O-])(=O)=O.[O-]S([O-])(=O)=O
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)